siRNA Details
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172282.2 | NP_758486.1 | 2 | 639 | ||
GenBank mRNA | AK033499.1 | - | 2 | 400 | ||
AK046065.1 | - | 2 | 429 | |||
AK049974.1 | - | 2 | 398 | |||
AK083321.1 | - | 2 | 463 | |||
AK088866.1 | - | 2 | 423 | |||
AK148252.1 | - | 2 | 442 | |||
BC039955.1 | - | 2 | 385 | 4236297 | ||
BC053487.1 | - | 2 | 414 |
