siRNA Details
Gene Symbol: Ube2q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180600.3 | NP_850931.2 | 4, 5 | 715 | ||
GenBank mRNA | AK042515.1 | - | 4, 5 | 716 | ||
AK076148.1 | - | 4, 5 | 578 | |||
AK083216.1 | - | 4, 5 | 708 | |||
AK150776.1 | - | 4, 5 | 540 | |||
BC029111.1 | - | 4, 5 | 544 | 2631258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
