siRNA Details
Gene Symbol: Nup214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172268.2 | NP_758472.2 | 19 | 2719 | ||
GenBank mRNA | AK122189.1 | - | 19 | 2688 | ||
BC039282.1 | - | 9 | 1076 | 4976266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
