siRNA Details
Gene Symbol: Sfswap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172276.3 | NP_758480.2 | 2, 3 | 534 | ||
GenBank mRNA | AK031872.1 | - | 2, 3 | 525 | ||
AK140369.1 | - | 2, 3 | 527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
