siRNA Details
Gene Symbol: 2210018M11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172280.2 | NP_758484.2 | 6 | 471 | ||
GenBank mRNA | AK032985.1 | - | 5 | 456 | ||
BC039956.1 | - | 6 | 462 | |||
BC066050.1 | - | 4 | 331 | |||
BC089304.1 | - | 6 | 471 | 6854147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
