siRNA Details
Gene Symbol: Cyp4a12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172306.2 | NP_758510.2 | 2 | 271 | ||
GenBank mRNA | AK098123.1 | - | 2 | 252 | ||
BB665127.1 | - | 2 | 273 | |||
BC060945.1 | - | 2 | 252 | 30304458 |
Gene Symbol: Cyp4a12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177406.3 | NP_803125.2 | 2 | 294 | ||
GenBank mRNA | AK144260.1 | - | 2 | 224 | ||
BC014721.1 | - | 2 | 240 | 4240359 | ||
BC025936.1 | - | 2 | 234 | 5097698 | ||
BC026582.1 | - | 2 | 240 | 4971335 | ||
BC031141.1 | - | 2 | 215 | 4972684 | ||
BC033924.1 | - | 2 | 224 | 5101821 | ||
CB955255.1 | - | 2 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
