siRNA Details
Gene Symbol: Mbtps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172307.3 | NP_758511.1 | 5, 6 | 797 | ||
GenBank mRNA | AK148325.1 | - | 5, 6 | 762 | ||
AK164486.1 | - | 5, 6 | 733 | |||
BC038343.1 | - | 5, 6 | 724 | 4460515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
