siRNA Details
Gene Symbol: Pddc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172116.4 | NP_742114.3 | 8 | 852 | ||
GenBank mRNA | AK081187.1 | - | 8 | 812 | ||
AK084273.1 | - | 8 | 808 | |||
AK140975.1 | - | 8 | 803 | |||
AK150323.1 | - | 6 | 1522 | |||
AK150354.1 | - | 6 | 1522 | |||
AK151259.1 | - | 6 | 1522 | |||
AK151794.1 | - | 6 | 1522 | |||
AK151959.1 | - | 6 | 1522 | |||
AK153110.1 | - | 6 | 1522 | |||
AK161205.1 | - | 7 | 1655 | |||
BC023835.1 | - | 8 | 822 | 5345186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
