siRNA Details
Gene Symbol: Mterf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013023.2 | NP_001013041.2 | 2 | 1308 | ||
NM_172135.2 | NP_742147.1 | 2 | 1289 | |||
GenBank mRNA | AY726770.2 | - | 2 | 1301 | ||
BC038058.1 | - | 2 | 1281 | 4207737 | ||
BC051251.1 | - | 2 | 1302 | 6490147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
