siRNA Details
Gene Symbol: Zdhhc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172151.4 | NP_742163.4 | 11 | 3058 | ||
GenBank mRNA | AK032069.1 | - | 2 | 1048 | ||
AY894892.1 | - | 11 | 3058 | |||
BC004683.1 | - | 1 | 869 | |||
BC055694.1 | - | 2 | 1355 | |||
BC083154.1 | - | 1 | 979 | |||
BC106166.1 | - | 1 | 961 |

More Information
Other Species Targeted

Gene Ontology Categories:
