siRNA Details
Gene Symbol: Zmynd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252587.2 | NP_001239516.1 | 12 | 2171 | ||
GenBank mRNA | AK017513.1 | - | 1 | 236 | ||
AK021201.1 | - | 1 | 287 | |||
BC020059.1 | - | 3 | 731 | |||
BC023300.1 | - | 12 | 1812 | |||
BC063267.1 | - | 11 | 1951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
