siRNA Details
Gene Symbol: Sfswap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172276.3 | NP_758480.2 | 2, 3 | 546 | ||
GenBank mRNA | AK031872.1 | - | 2, 3 | 537 | ||
AK088182.1 | - | 2, 3 | 490 | |||
AK140369.1 | - | 2, 3 | 539 | |||
BC057314.1 | - | 2, 3 | 2476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
