siRNA Details
Gene Symbol: Plcg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172285.1 | NP_758489.1 | 30 | 3560 | ||
GenBank mRNA | AK142173.1 | - | 30 | 3563 | ||
AK170430.1 | - | 30 | 3571 | |||
BC019654.1 | - | 17 | 2094 | |||
BC023877.1 | - | 30 | 3560 | 5354925 | ||
BC037700.1 | - | 12 | 1352 | |||
BC049273.1 | - | 13 | 1486 |
