siRNA Details
Gene Symbol: Tex10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172304.3 | NP_758508.2 | 8 | 1919 | ||
GenBank mRNA | AK132870.1 | - | 8 | 1920 | ||
AK141288.1 | - | 7 | 1698 | |||
BC006867.1 | - | 5 | 1166 | |||
BC151176.1 | - | 7 | 1670 | 9088065 | ||
BC151177.1 | - | 7 | 1670 | 9088066 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
