siRNA Details
Gene Symbol: Tarsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172310.2 | NP_758514.2 | 19 | 2293 | ||
GenBank mRNA | AK040927.1 | - | 19 | 2266 | ||
AK079416.1 | - | 19 | 2275 | |||
BC039225.1 | - | 19 | 2277 | 4458085 |
Gene Symbol: Tm2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077858.1 | - | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
