siRNA Details
Gene Symbol: RRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033.4 | NP_001024.1 | 8 | 1024 | ||
NM_001318064.1 | NP_001304993.1 | 7 | 846 | |||
NM_001318065.1 | NP_001304994.1 | 2 | 329 | |||
XM_011520278.1 | XP_011518580.1 | 2 | 324 | |||
GenBank mRNA | AK222786.1 | - | 8 | 904 | ||
AK297988.1 | - | 8 | 769 | |||
AK299629.1 | - | 6 | 661 | |||
AK301891.1 | - | 2 | 164 | |||
AK301922.1 | - | 2 | 164 | |||
AK303394.1 | - | 7 | 690 | |||
AK307950.1 | - | 9 | 809 | |||
AK311593.1 | - | 2 | 329 | |||
BC006498.2 | - | 8 | 855 | 2989344 | ||
X59543.1 | - | 8 | 883 | |||
X59617.1 | - | 8 | 877 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
