siRNA Details
Gene Symbol: RRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034.3 | NP_001025.1 | 4 | 437 | ||
NM_001165931.1 | NP_001159403.1 | 4 | 605 | |||
GenBank mRNA | AK092671.1 | - | 4 | 406 | ||
AK123010.1 | - | 4 | 373 | |||
AK313719.1 | - | 4 | 434 | |||
BC001886.1 | - | 4 | 418 | 3528619 | ||
BC030154.1 | - | 4 | 423 | 4858919 | ||
S40301.1 | - | 4 | 568 | |||
X59618.1 | - | 4 | 568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
