siRNA Details
Gene Symbol: Fmnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172409.2 | NP_765997.2 | 22, 23 | 3124 | ||
GenBank mRNA | AF513716.1 | - | 11, 12 | 1521 | ||
AK080318.1 | - | 2, 3 | 299 | |||
AK173293.1 | - | 5, 6 | 471 | |||
BC048004.1 | - | 7, 8 | 1012 | |||
BC064731.1 | - | 2, 3 | 281 |

More Information
Other Species Targeted

Gene Ontology Categories:
