siRNA Details
Gene Symbol: Sphkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172430.3 | NP_766018.3 | 2 | 258 | ||
GenBank mRNA | AK020841.1 | - | 3 | 233 | ||
AK034183.1 | - | 3 | 301 | |||
AK038638.1 | - | 3 | 303 | |||
BY769819.1 | - | 2 | 260 | |||
BY774616.1 | - | 2 | 259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
