siRNA Details
Gene Symbol: P2ry10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172435.3 | NP_766023.1 | 3 | 493 | ||
GenBank mRNA | AK020001.1 | - | 3 | 371 | ||
AK038124.1 | - | 2 | 420 | |||
AK041351.1 | - | 3 | 509 | |||
AK088234.1 | - | 3 | 494 | |||
BC053707.1 | - | 3 | 324 | 30072257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
