siRNA Details
Gene Symbol: Shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290684.1 | NP_001277613.1 | 7 | 3323 | ||
NM_001290685.1 | NP_001277614.1 | 3 | 648 | |||
NM_001290686.1 | NP_001277615.1 | 3 | 455 | |||
NM_001290687.1 | NP_001277616.1 | 3 | 637 | |||
NM_172441.3 | NP_766029.2 | 7 | 3341 | |||
GenBank mRNA | AK029338.1 | - | 3 | 329 | ||
AK032256.1 | - | 3 | 330 | |||
AK049850.1 | - | 3 | 387 | |||
BC092373.1 | - | 3 | 643 | |||
CF534250.1 | - | 3 | 598 | |||
CK635066.1 | - | 4 | 606 | |||
CO431718.1 | - | 3 | 637 | |||
DQ358971.1 | - | 7 | 3304 | |||
EF071946.1 | - | 7 | 3341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
