siRNA Details
Gene Symbol: Mob3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172457.2 | NP_766045.2 | 4 | 1847 | ||
GenBank mRNA | AK030481.1 | - | 4 | 1884 | ||
AK041672.1 | - | 751 | ||||
AK164429.1 | - | 1140 | ||||
AK170394.1 | - | 4 | 1858 | |||
AK171197.1 | - | 1871 | ||||
BC051170.1 | - | 3 | 1836 | |||
BC058238.1 | - | 4 | 1847 | 6307543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
