siRNA Details
Gene Symbol: Zfp871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172458.3 | NP_766046.3 | 2 | 257 | ||
GenBank mRNA | AI119099.1 | - | 2 | 257 | ||
AK029041.1 | - | 2 | 245 | |||
AK122579.1 | - | 2 | 220 | |||
BC043671.1 | - | 2 | 372 | 5040597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
