siRNA Details
Gene Symbol: Dip2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036188.1 | - | 2, 3 | 212 | ||
BC089303.1 | - | 3, 4 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
