siRNA Details
Gene Symbol: Fmnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172409.2 | NP_765997.2 | 24 | 3238 | ||
GenBank mRNA | AF513716.1 | - | 13 | 1635 | ||
AK080318.1 | - | 4 | 413 | |||
AK173293.1 | - | 7 | 585 | |||
BC048004.1 | - | 9 | 1126 | |||
BC064731.1 | - | 4 | 395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
