siRNA Details
Gene Symbol: A630033H20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122595.1 | NP_001116067.1 | 2 | 735 | ||
NM_001122596.1 | NP_001116068.1 | 2 | 782 | |||
NM_175442.4 | NP_780651.1 | 2 | 746 | |||
GenBank mRNA | AK041740.1 | - | 2 | 747 |
Gene Symbol: P2ry10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172435.3 | NP_766023.1 | 3 | 681 | ||
GenBank mRNA | AK020001.1 | - | 3 | 560 | ||
AK038124.1 | - | 2 | 608 | |||
AK041351.1 | - | 3 | 697 | |||
AK088234.1 | - | 3 | 682 | |||
BC053707.1 | - | 3 | 512 | 30072257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
