siRNA Details
Gene Symbol: Dip2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081426.2 | NP_001074895.2 | 24 | 3387 | ||
GenBank mRNA | AK036188.1 | - | 3 | 242 | ||
AK173066.1 | - | 16 | 1799 | |||
BC056940.1 | - | 7 | 851 | |||
BC066797.1 | - | 4 | 433 | |||
BC089303.1 | - | 4 | 411 | |||
BC137787.1 | - | 24 | 2979 | 8860806 | ||
BC144909.1 | - | 23 | 2892 | 9053457 | ||
BC144910.1 | - | 24 | 2979 | 9053458 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
