siRNA Details
Gene Symbol: Aim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172393.2 | NP_765981.2 | 14 | 4975 | ||
GenBank mRNA | AK041082.1 | - | 13 | 1591 | ||
AK088102.1 | - | 13 | 1647 | |||
BC035551.1 | - | 13 | 2127 | |||
BC059272.1 | - | 13 | 2132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
