siRNA Details
Gene Symbol: Fam175a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172405.3 | NP_765993.1 | 9 | 1580 | ||
GenBank mRNA | AK051816.1 | - | 9 | 1581 | ||
AK167540.1 | - | 9 | 1611 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
