siRNA Details
Gene Symbol: Fmnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172409.2 | NP_765997.2 | 27 | 3964 | ||
GenBank mRNA | AK080318.1 | - | 7 | 1139 | ||
AK173293.1 | - | 10 | 1358 | |||
BC048004.1 | - | 12 | 1900 | |||
BC064731.1 | - | 6 | 1074 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
