siRNA Details
Gene Symbol: Olfr112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013575.4 | NP_001013593.2 | 3 | 3017 | ||
GenBank mRNA | AK029968.1 | - | 3 | 3015 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
