siRNA Details
Gene Symbol: Shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290684.1 | NP_001277613.1 | 10 | 4606 | ||
NM_001290685.1 | NP_001277614.1 | 6 | 1931 | |||
NM_001290686.1 | NP_001277615.1 | 6 | 1738 | |||
NM_001290687.1 | NP_001277616.1 | 6 | 1920 | |||
NM_172441.3 | NP_766029.2 | 10 | 4624 | |||
GenBank mRNA | AK029338.1 | - | 6 | 1612 | ||
AK032256.1 | - | 6 | 1613 | |||
AK049833.1 | - | 2 | 592 | |||
AK049850.1 | - | 6 | 1670 | |||
BC092373.1 | - | 6 | 1926 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
