siRNA Details
Gene Symbol: Dip2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081426.2 | NP_001074895.2 | 37 | 5795 | ||
GenBank mRNA | AK036188.1 | - | 16 | 2650 | ||
AK173066.1 | - | 29 | 4207 | |||
BC056940.1 | - | 20 | 3260 | |||
BC066797.1 | - | 17 | 2841 | |||
BC089303.1 | - | 17 | 2819 |

More Information
Other Species Targeted

Gene Ontology Categories:
