siRNA Details
Gene Symbol: Frmd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177843.1 | NP_001171314.1 | 24 | 4754 | ||
NM_001177844.1 | NP_001171315.1 | 12 | 3810 | |||
NM_172475.3 | NP_766063.3 | 23 | 4312 | |||
GenBank mRNA | AB093292.1 | - | 10 | 3078 | ||
AK082365.1 | - | 24 | 4331 | |||
AK142412.1 | - | 1 | 782 | |||
AK148414.1 | - | 13 | 3811 | |||
AK171532.1 | - | 12 | 3738 | |||
BC058672.1 | - | 23 | 4303 | 6830456 | ||
BC082808.1 | - | 24 | 4759 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
