siRNA Details
Gene Symbol: Zfp677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172486.2 | NP_766074.2 | 4 | 1321 | ||
GenBank mRNA | AK043955.1 | - | 4 | 1323 | ||
BC057947.1 | - | 4 | 1299 | 5344769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
