siRNA Details
Gene Symbol: Spg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145531.2 | NP_663506.2 | 2 | 287 | ||
GenBank mRNA | AK039286.1 | - | 2 | 238 | ||
AK049236.1 | - | 2 | 279 | |||
AK049617.1 | - | 2 | 198 | |||
AK147500.1 | - | 2 | 288 |

More Information
Other Species Targeted

Gene Ontology Categories:
