siRNA Details
Gene Symbol: Baz1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013815.2 | NP_038843.2 | 18 | 2829 | ||
GenBank mRNA | AK047427.1 | - | 3 | 637 | ||
AK132694.1 | - | 1 | 24 | |||
BC065123.1 | - | 4 | 645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
