siRNA Details
Gene Symbol: Unc13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029873.2 | NP_001025044.2 | 44 | 7733 | ||
GenBank mRNA | AK039579.1 | - | 40 | |||
AK083196.1 | - | 1 | 2237 | |||
AK141188.1 | - | 2817 | ||||
AK147408.1 | - | 39 | 7246 | |||
BC058348.1 | - | 1 | 671 | |||
CJ250255.1 | - | 1 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
