siRNA Details
Gene Symbol: Hjurp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198652.2 | NP_941054.2 | 7 | 581 | ||
GenBank mRNA | AB162219.1 | - | 7 | 471 | ||
BC057309.1 | - | 7 | 522 | 6826508 |
Gene Symbol: 6430706D22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062125.1 | - | 2 | 112 | 6826662 |
Gene Symbol: A730008H23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172505.4 | NP_766093.1 | 4 | 332 | ||
GenBank mRNA | AK042589.1 | - | 4 | 332 | ||
AK138428.1 | - | 4 | 295 | |||
BC131919.1 | - | 4 | 300 | 40130196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
