siRNA Details
Gene Symbol: Spg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145531.2 | NP_663506.2 | 2, 3 | 450 | ||
GenBank mRNA | AK039286.1 | - | 2, 3 | 401 | ||
AK049236.1 | - | 2, 3 | 442 | |||
AK049617.1 | - | 2, 3 | 361 | |||
AK147500.1 | - | 2, 3 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
