siRNA Details
Gene Symbol: Adamtsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029967.3 | NP_084243.3 | 18, 19 | 3777 | ||
GenBank mRNA | AK031246.1 | - | 1, 2 | 380 | ||
AK132420.1 | - | 6, 7 | 2217 | |||
BC116295.1 | - | 1, 2 | 371 | |||
BC116296.1 | - | 1, 2 | 371 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008763845.
Gene Ontology Categories:
