siRNA Details
Gene Symbol: Fam126b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172513.3 | NP_766101.3 | 12 | 2000 | ||
GenBank mRNA | AK052638.1 | - | 12 | 1927 | ||
AK144602.1 | - | 13 | 2105 | |||
AK168372.1 | - | 12 | 1990 | |||
BC049806.1 | - | 12 | 1936 |

More Information
Other Species Targeted

Gene Ontology Categories:
