siRNA Details
Gene Symbol: Spg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145531.2 | NP_663506.2 | 5 | 992 | ||
GenBank mRNA | AK039286.1 | - | 5 | 943 | ||
AK049236.1 | - | 5 | 984 | |||
AK049617.1 | - | 5 | 903 | |||
AK147500.1 | - | 5 | 993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
