siRNA Details
Gene Symbol: Ehmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012518.3 | NP_001012536.2 | 27 | 4849 | ||
NM_001109686.2 | NP_001103156.1 | 26 | 4690 | |||
NM_001109687.2 | NP_001103157.1 | 26 | 4705 | |||
NM_172545.4 | NP_766133.2 | 27 | 4828 | |||
GenBank mRNA | AK020308.1 | - | 1 | 754 | ||
AK082062.1 | - | 27 | 4840 | |||
AK149125.1 | - | 7 | 1772 | |||
AV572776.1 | - | 1 | 122 | |||
DV649682.1 | - | 1 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
