siRNA Details
Gene Symbol: Adamts10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172619.3 | NP_766207.2 | 5 | 779 | ||
GenBank mRNA | AK034565.1 | - | 5 | 754 | ||
AK042525.1 | - | 5 | 767 | |||
AK076334.1 | - | 5 | 753 | |||
BC056427.1 | - | 5 | 749 | |||
BC082773.1 | - | 5 | 771 | |||
BC100558.1 | - | 4 | 533 | |||
BC145353.1 | - | 4 | 686 | |||
BC145354.1 | - | 4 | 686 | |||
BC145355.1 | - | 4 | 686 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
