siRNA Details
Gene Symbol: 4932438A13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172679.2 | NP_766267.2 | 7 | 746 | ||
GenBank mRNA | AK028716.1 | - | 6 | 422 | ||
AK030086.1 | - | 7 | 733 | |||
AK041411.1 | - | 7 | 701 | |||
AK148181.1 | - | 4 | 256 | |||
AK172264.1 | - | 7 | 681 | |||
AK220265.1 | - | 7 | 696 | |||
BC064036.1 | - | 5 | 391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
