siRNA Details
Gene Symbol: 4932438A13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172679.2 | NP_766267.2 | 76 | 13263 | ||
GenBank mRNA | AK041642.1 | - | 3 | 347 | ||
AK052702.1 | - | 8 | 1081 | |||
AK164468.1 | - | 2 | 861 | |||
BC027125.1 | - | 3 | 295 | |||
BC079623.1 | - | 17 | 2633 |

More Information
Other Species Targeted

Gene Ontology Categories:
