siRNA Details
Gene Symbol: N28178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172690.2 | NP_766278.1 | 4 | 399 | ||
GenBank mRNA | AK038528.1 | - | 3 | 253 | ||
AK046283.1 | - | 4 | 390 | |||
AK122428.1 | - | 2 | 322 | |||
BC057092.1 | - | 4 | 399 | 5703966 |
