siRNA Details
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005787.1 | NP_001005787.1 | 2 | 389 | ||
NM_007704.2 | NP_031730.1 | 2 | 389 | |||
NM_172696.2 | NP_766284.2 | 28 | 3839 | |||
GenBank mRNA | AF060539.1 | - | 2 | 289 | ||
AK077268.1 | - | 15 | 2121 | |||
AK147866.1 | - | 28 | 3788 | |||
BC037607.1 | - | 2 | 61 | |||
BC057124.1 | - | 2 | 68 | |||
BC082787.1 | - | 17 | 2232 | |||
BC141404.1 | - | 28 | 3815 | 9056017 |

More Information
Other Species Targeted

Gene Ontology Categories:
