siRNA Details
Gene Symbol: Atp13a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164612.1 | NP_001158084.1 | 11 | 1387 | ||
NM_001164613.1 | NP_001158085.1 | 12 | 1440 | |||
GenBank mRNA | AK028776.1 | - | 12 | 1442 | ||
AK029303.1 | - | 10 | 1131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
